3,4-Dimethyl-2-(1H-pyrazol-1-yl)benzaldehyde
Supplier: Matrix Scientific
053710-500MG
053710-2.5G
101904-710EA
364.62
USD
101904-710
101904-712
3,4-Dimethyl-2-(1H-pyrazol-1-yl)benzaldehyde
3,4-Dimethyl-2-(1H-pyrazol-1-yl)benzaldehyde
Hazard: Irritant
Formula:
C₁₂H₁₂N₂O |
MDL Number:
MFCD09054726 CAS Number: 1214622-41-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...