3,5-Dimethyl-2-(1H-pyrazol-1-yl)benzaldehyde
Supplier: Matrix Scientific
053707-500MG
053707-2.5G
101904-698EA
364.62
USD
101904-698
101904-700
3,5-Dimethyl-2-(1H-pyrazol-1-yl)benzaldehyde
3,5-Dimethyl-2-(1H-pyrazol-1-yl)benzaldehyde
Hazard: Irritant
Formula:
C₁₂H₁₂N₂O |
CAS Number:
1015845-87-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...