5-Fluoro-2-(1H-pyrazol-1-yl)benzaldehyde
Supplier: Matrix Scientific
053705-500MG
053705-2.5G
101904-690EA
158.46
USD
101904-690
101904-692
5-Fluoro-2-(1H-pyrazol-1-yl)benzaldehyde
5-Fluoro-2-(1H-pyrazol-1-yl)benzaldehyde
Hazard: Irritant
Formula:
C₁₀H₇FN₂O |
MDL Number:
MFCD08059861 CAS Number: 1015845-84-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...