3-(4-Ethoxybenzyl)piperidine

Supplier: Matrix Scientific

053703-500MG 053703-2.5G
101904-682EA 149.23 USD
101904-682 101904-684
3-(4-Ethoxybenzyl)piperidine
3-(4-Ethoxybenzyl)piperidine

Hazard: Irritant

Formula: C₁₄H₂₁NO
MDL Number: MFCD05863595
CAS Number: 955315-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR