3-(3,5-Dimethoxybenzyl)piperidine

Supplier: Matrix Scientific

053702-500MG 053702-2.5G
101904-678EA 364.62 USD
101904-678 101904-680
3-(3,5-Dimethoxybenzyl)piperidine
3-(3,5-Dimethoxybenzyl)piperidine

Hazard: Irritant

Formula: C₁₄H₂₁NO₂
MDL Number: MFCD13562911
CAS Number: 1220035-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR