3-(2-Methoxybenzyl)piperidine
Supplier: Matrix Scientific
053701-500MG
053701-2.5G
101904-674EA
166.15
USD
101904-674
101904-676
3-(2-Methoxybenzyl)piperidine
3-(2-Methoxybenzyl)piperidine
Hazard: Irritant
Formula:
C₁₃H₁₉NO MW: 205.3 g/mol |
MDL Number:
MFCD05863594 CAS Number: 420137-10-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...