3-Propylpiperidine

Supplier: Matrix Scientific

053690-500MG 053690-2.5G
101904-638EA 364.62 USD
101904-638 101904-640
3-Propylpiperidine
3-Propylpiperidine

Hazard: Irritant

Formula: C₈H₁₇N
MDL Number: MFCD01700132
CAS Number: 13603-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR