3-[2-(2-Methoxyethoxy)ethyl]piperidine

Supplier: Matrix Scientific

053687-500MG 053687-2.5G
101904-626EA 364.62 USD
101904-626 101904-628
3-[2-(2-Methoxyethoxy)ethyl]piperidine
3-[2-(2-Methoxyethoxy)ethyl]piperidine

Hazard: Irritant

Formula: C₁₀H₂₁NO₂
MDL Number: MFCD13562903
CAS Number: 1220024-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR