3-(2-Ethoxyethyl)piperidine
Supplier: Matrix Scientific
053686-500MG
053686-2.5G
101904-622EA
364.62
USD
101904-622
101904-624
3-(2-Ethoxyethyl)piperidine
3-(2-Ethoxyethyl)piperidine
Hazard: Irritant
Formula:
C₉H₁₉NO |
MDL Number:
MFCD08687705 CAS Number: 946682-08-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...