3-(2-Methoxyethyl)piperidine

Supplier: Matrix Scientific

053685-500MG 053685-2.5G
101904-618EA 364.62 USD
101904-618 101904-620
3-(2-Methoxyethyl)piperidine
3-(2-Methoxyethyl)piperidine

Hazard: Irritant

Formula: C₈H₁₇NO
MW: 143.23 g/mol
MDL Number: MFCD08688047
CAS Number: 946715-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR