3-(2-Phenoxyethyl)piperidine hydrochloride

Supplier: Matrix Scientific

053684-500MG 053684-2.5G
101904-614EA 364.62 USD
101904-614 101904-616
3-(2-Phenoxyethyl)piperidine hydrochloride
3-(2-Phenoxyethyl)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀ClNO
MDL Number: MFCD12026509
CAS Number: 1185298-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR