3-Isobutylpiperidine

Supplier: Matrix Scientific

053680-500MG 053680-2.5G
101904-598EA 461.54 USD
101904-598 101904-600
3-Isobutylpiperidine
3-Isobutylpiperidine

Hazard: Irritant

Formula: C₉H₁₉N
MDL Number: MFCD07383808
CAS Number: 956429-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR