3-Methylpyrrolidine hydrochloride
Supplier: Matrix Scientific
053678-500MG
053678-2.5G
101904-590EA
153.85
USD
101904-590
101904-592
3-Methylpyrrolidine hydrochloride
3-Methylpyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₅H₁₁N·ClH MW: 121.608 g/mol |
MDL Number:
MFCD06796630 CAS Number: 34375-89-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...