3-(4-Ethoxybenzyl)pyrrolidine

Supplier: Matrix Scientific

053677-500MG 053677-2.5G
101904-586EA 364.62 USD
101904-586 101904-588
3-(4-Ethoxybenzyl)pyrrolidine
3-(4-Ethoxybenzyl)pyrrolidine

Hazard: Irritant

Formula: C₁₃H₁₉NO
MDL Number: MFCD07383264
CAS Number: 1219977-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR