3-(3,5-Dimethoxybenzyl)pyrrolidine hydrochloride
Supplier: Matrix Scientific
053676-500MG
053676-2.5G
101904-582EA
364.62
USD
101904-582
101904-584
3-(3,5-Dimethoxybenzyl)pyrrolidine hydrochloride
3-(3,5-Dimethoxybenzyl)pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₁₃H₂₀ClNO₂ |
MDL Number:
MFCD21605759 CAS Number: 1361115-33-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...