3-[2-(2-Ethoxyethoxy)ethyl]pyrrolidine
Supplier: Matrix Scientific
053669-500MG
053669-2.5G
101904-554EA
166.15
USD
101904-554
101904-556
3-[2-(2-Ethoxyethoxy)ethyl]pyrrolidine
3-[2-(2-Ethoxyethoxy)ethyl]pyrrolidine
Hazard: Irritant
Formula:
C₁₀H₂₁NO₂ |
MDL Number:
MFCD13562896 CAS Number: 1219977-22-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...