3-(4-Methylpentyl)pyrrolidine
Supplier: Matrix Scientific
053662-500MG
053662-2.5G
101904-534EA
364.62
USD
101904-534
101904-536
3-(4-Methylpentyl)pyrrolidine
3-(4-Methylpentyl)pyrrolidine
Hazard: Irritant
Formula:
C₁₀H₂₁N |
MDL Number:
MFCD13562889 CAS Number: 1220021-17-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...