3-(Cyclopropylmethyl)pyrrolidine hydrochloride
Supplier: Matrix Scientific
053660-500MG
053660-2.5G
101904-526EA
364.62
USD
101904-526
101904-528
3-(Cyclopropylmethyl)pyrrolidine hydrochloride
3-(Cyclopropylmethyl)pyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₈H₁₆ClN |
MDL Number:
MFCD27579022 CAS Number: 1707604-64-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...