3-(2-Ethoxyethyl)pyrrolidine

Supplier: Matrix Scientific

053653-500MG 053653-2.5G
101904-498EA 384.62 USD
101904-498 101904-500
3-(2-Ethoxyethyl)pyrrolidine
3-(2-Ethoxyethyl)pyrrolidine

Hazard: Irritant

Formula: C₈H₁₇NO
MDL Number: MFCD13562883
CAS Number: 1220038-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR