3-(2-Phenoxyethyl)pyrrolidine

Supplier: Matrix Scientific

053651-500MG 053651-2.5G
101904-490EA 461.54 USD
101904-490 101904-492
3-(2-Phenoxyethyl)pyrrolidine
3-(2-Phenoxyethyl)pyrrolidine

Hazard: Irritant

Formula: C₁₂H₁₇NO
MDL Number: MFCD13562881
CAS Number: 1220035-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR