3-(2-Ethylhexyl)pyrrolidine
Supplier: Matrix Scientific
053649-500MG
053649-2.5G
101904-482EA
364.62
USD
101904-482
101904-484
3-(2-Ethylhexyl)pyrrolidine
3-(2-Ethylhexyl)pyrrolidine
Hazard: Irritant
Formula:
C₁₂H₂₅N |
MDL Number:
MFCD13562880 CAS Number: 1220019-23-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...