3-Isobutylpyrrolidine hydrochloride
Supplier: Matrix Scientific
053646-500MG
053646-2.5G
101904-470EA
461.54
USD
101904-470
101904-472
3-Isobutylpyrrolidine hydrochloride
3-Isobutylpyrrolidine hydrochloride
Hazard: Irritant
Formula:
C₈H₁₈ClN MW: 163.69 g/mol |
MDL Number:
MFCD27920997 CAS Number: 959238-03-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...