3-Benzylpyrrolidine

Supplier: Matrix Scientific

053644-500MG 053644-2.5G
101904-462EA 364.62 USD
101904-462 101904-464
3-Benzylpyrrolidine
3-Benzylpyrrolidine
Formula: C₁₁H₁₅N
MDL Number: MFCD01862537
CAS Number: 170304-83-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR