2-(Cyclopropylmethoxy)-5-propylbenzaldehyde
Supplier: Matrix Scientific
053643-500MG
053643-2.5G
101904-458EA
364.62
USD
101904-458
101904-460
2-(Cyclopropylmethoxy)-5-propylbenzaldehyde
2-(Cyclopropylmethoxy)-5-propylbenzaldehyde
Hazard: Irritant
Formula:
C₁₄H₁₈O₂ |
MDL Number:
MFCD06246807 CAS Number: 883528-71-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...