2-Propoxy-5-propylbenzaldehyde

Supplier: Matrix Scientific

053642-500MG 053642-2.5G
101904-454EA 364.62 USD
101904-454 101904-456
2-Propoxy-5-propylbenzaldehyde
2-Propoxy-5-propylbenzaldehyde

Hazard: Irritant

Formula: C₁₃H₁₈O₂
MDL Number: MFCD06248743
CAS Number: 883533-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR