2-Methoxy-5-propylbenzaldehyde

Supplier: Matrix Scientific

053640-500MG 053640-2.5G
101904-446EA 166.15 USD
101904-446 101904-448
2-Methoxy-5-propylbenzaldehyde
2-Methoxy-5-propylbenzaldehyde

Hazard: Irritant

Formula: C₁₁H₁₄O₂
Boiling Pt: 151 °C (16 Torr)
Density: 1.045
MDL Number: MFCD06248102
CAS Number: 145742-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR