3-Chloro-5-ethyl-2-propoxybenzaldehyde
Supplier: Matrix Scientific
053638-500MG
053638-2.5G
101904-438EA
364.62
USD
101904-438
101904-440
3-Chloro-5-ethyl-2-propoxybenzaldehyde
3-Chloro-5-ethyl-2-propoxybenzaldehyde
Hazard: Irritant
Formula:
C₁₂H₁₅ClO₂ |
MDL Number:
MFCD06246204 CAS Number: 883523-24-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...