3-Chloro-2-ethoxy-5-ethylbenzaldehyde
Supplier: Matrix Scientific
053637-500MG
053637-2.5G
101904-434EA
364.62
USD
101904-434
101904-436
3-Chloro-2-ethoxy-5-ethylbenzaldehyde
3-Chloro-2-ethoxy-5-ethylbenzaldehyde
Hazard: Irritant
Formula:
C₁₁H₁₃ClO₂ |
MDL Number:
MFCD06246177 CAS Number: 883521-52-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...