3-Chloro-5-ethyl-2-methoxybenzaldehyde
Supplier: Matrix Scientific
053636-500MG
053636-2.5G
101904-430EA
746.15
USD
101904-430
101904-432
3-Chloro-5-ethyl-2-methoxybenzaldehyde
3-Chloro-5-ethyl-2-methoxybenzaldehyde
Hazard: Irritant
Formula:
C₁₀H₁₁ClO₂ |
MDL Number:
MFCD06246202 CAS Number: 883523-11-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...