3-Chloro-2-(cyclopropylmethoxy)-5-methylbenzaldehyde

Supplier: Matrix Scientific

053635-500MG 053635-2.5G
101904-426EA 364.62 USD
101904-426 101904-428
3-Chloro-2-(cyclopropylmethoxy)-5-methylbenzaldehyde
3-Chloro-2-(cyclopropylmethoxy)-5-methylbenzaldehyde

Hazard: Irritant

Formula: C₁₂H₁₃ClO₂
MDL Number: MFCD06246153
CAS Number: 883520-21-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR