3-Chloro-5-methyl-2-propoxybenzaldehyde
Supplier: Matrix Scientific
053634-500MG
053634-2.5G
101904-422EA
364.62
USD
101904-422
101904-424
3-Chloro-5-methyl-2-propoxybenzaldehyde
3-Chloro-5-methyl-2-propoxybenzaldehyde
Hazard: Irritant
Formula:
C₁₁H₁₃ClO₂ |
MDL Number:
MFCD06246360 CAS Number: 883517-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...