3-Chloro-2-ethoxy-5-methylbenzaldehyde
Supplier: Matrix Scientific
053633-500MG
053633-2.5G
101904-418EA
364.62
USD
101904-418
101904-420
3-Chloro-2-ethoxy-5-methylbenzaldehyde
3-Chloro-2-ethoxy-5-methylbenzaldehyde
Hazard: Irritant
Formula:
C₁₀H₁₁ClO₂ MW: 198.65 g/mol |
MDL Number:
MFCD06246189 CAS Number: 883522-32-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...