3-Chloro-2-methoxy-5-methylbenzaldehyde
Supplier: Matrix Scientific
053632-500MG
053632-2.5G
101904-414EA
364.62
USD
101904-414
101904-416
3-Chloro-2-methoxy-5-methylbenzaldehyde
3-Chloro-2-methoxy-5-methylbenzaldehyde
Hazard: Irritant
Formula:
C₉H₉ClO₂ |
MDL Number:
MFCD06246301 CAS Number: 883529-23-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...