5-Ethyl-2-propoxybenzaldehyde
Supplier: Matrix Scientific
053630-500MG
053630-2.5G
101904-406EA
166.15
USD
101904-406
101904-408
5-Ethyl-2-propoxybenzaldehyde
5-Ethyl-2-propoxybenzaldehyde
Hazard: Irritant
Formula:
C₁₂H₁₆O₂ |
MDL Number:
MFCD06247484 CAS Number: 883528-61-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...