2-Ethoxy-5-ethylbenzaldehyde

Supplier: Matrix Scientific

053629-500MG 053629-2.5G
101904-402EA 461.54 USD
101904-402 101904-404
2-Ethoxy-5-ethylbenzaldehyde
2-Ethoxy-5-ethylbenzaldehyde

Hazard: Irritant

Formula: C₁₁H₁₄O₂
MDL Number: MFCD06247338
CAS Number: 883536-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR