5-Ethyl-2-methoxybenzaldehyde

Supplier: Matrix Scientific

053628-500MG 053628-2.5G
101904-398EA 384.62 USD
101904-398 101904-400
5-Ethyl-2-methoxybenzaldehyde
5-Ethyl-2-methoxybenzaldehyde

Hazard: Irritant

Formula: C₁₀H₁₂O₂
MDL Number: MFCD06247461
CAS Number: 85944-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR