2-(Cyclopropylmethoxy)-5-methylbenzaldehyde
Supplier: Matrix Scientific
053627-500MG
053627-2.5G
101904-394EA
384.62
USD
101904-394
101904-396
2-(Cyclopropylmethoxy)-5-methylbenzaldehyde
2-(Cyclopropylmethoxy)-5-methylbenzaldehyde
Hazard: Irritant
Formula:
C₁₂H₁₄O₂ |
MDL Number:
MFCD06246774 CAS Number: 883526-60-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...