5-Methyl-2-propoxybenzaldehyde

Supplier: Matrix Scientific

053626-500MG 053626-2.5G
101904-390EA 226.15 USD
101904-390 101904-392
5-Methyl-2-propoxybenzaldehyde
5-Methyl-2-propoxybenzaldehyde

Hazard: Irritant

Formula: C₁₁H₁₄O₂
MDL Number: MFCD06248399
CAS Number: 883531-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR