2-Ethoxy-5-methylbenzaldehyde
Supplier: Matrix Scientific
053625-500MG
053625-2.5G
101904-386EA
226.15
USD
101904-386
101904-388
2-Ethoxy-5-methylbenzaldehyde
2-Ethoxy-5-methylbenzaldehyde
Hazard: Irritant
Formula:
C₁₀H₁₂O₂ |
MDL Number:
MFCD06247359 CAS Number: 116529-99-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...