5-(sec-Butyl)-2-(cyclopropylmethoxy)benzaldehyde

Supplier: Matrix Scientific

053624-500MG 053624-2.5G
101904-382EA 364.62 USD
101904-382 101904-384
5-(sec-Butyl)-2-(cyclopropylmethoxy)benzaldehyde
5-(sec-Butyl)-2-(cyclopropylmethoxy)benzaldehyde

Hazard: Irritant

Formula: C₁₅H₂₀O₂
MDL Number: MFCD06246071
CAS Number: 883514-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR