5-(sec-Butyl)-2-propoxybenzaldehyde

Supplier: Matrix Scientific

053623-500MG 053623-2.5G
101904-378EA 364.62 USD
101904-378 101904-380
5-(sec-Butyl)-2-propoxybenzaldehyde
5-(sec-Butyl)-2-propoxybenzaldehyde

Hazard: Irritant

Formula: C₁₄H₂₀O₂
MDL Number: MFCD06246096
CAS Number: 883515-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR