5-(sec-Butyl)-2-methoxybenzaldehyde

Supplier: Matrix Scientific

053621-500MG 053621-2.5G
101904-370EA 384.62 USD
101904-370 101904-372
5-(sec-Butyl)-2-methoxybenzaldehyde
5-(sec-Butyl)-2-methoxybenzaldehyde

Hazard: Irritant

Formula: C₁₂H₁₆O₂
MDL Number: MFCD06246085
CAS Number: 145742-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR