2-(Cyclopropylmethoxy)-5-isopropylbenzaldehyde

Supplier: Matrix Scientific

053620-500MG 053620-2.5G
101904-366EA 364.62 USD
101904-366 101904-368
2-(Cyclopropylmethoxy)-5-isopropylbenzaldehyde
2-(Cyclopropylmethoxy)-5-isopropylbenzaldehyde

Hazard: Irritant

Formula: C₁₄H₁₈O₂
MDL Number: MFCD06246764
CAS Number: 883525-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR