2-Ethoxy-5-isopropylbenzaldehyde
Supplier: Matrix Scientific
053618-500MG
053618-2.5G
101904-358EA
384.62
USD
101904-358
101904-360
2-Ethoxy-5-isopropylbenzaldehyde
2-Ethoxy-5-isopropylbenzaldehyde
Hazard: Irritant
Formula:
C₁₂H₁₆O₂ |
MDL Number:
MFCD03093066 CAS Number: 883537-51-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...