5-(tert-Butyl)-2-(cyclopropylmethoxy)benzaldehyde
Supplier: Matrix Scientific
053617-500MG
053617-2.5G
101904-354EA
364.62
USD
101904-354
101904-356
5-(tert-Butyl)-2-(cyclopropylmethoxy)benzaldehyde
5-(tert-Butyl)-2-(cyclopropylmethoxy)benzaldehyde
Hazard: Irritant
Formula:
C₁₅H₂₀O₂ |
MDL Number:
MFCD06246073 CAS Number: 883514-34-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...