5-(tert-Butyl)-2-propoxybenzaldehyde
Supplier: Matrix Scientific
053616-500MG
053616-2.5G
101904-350EA
364.62
USD
101904-350
101904-352
5-(tert-Butyl)-2-propoxybenzaldehyde
5-(tert-Butyl)-2-propoxybenzaldehyde
Hazard: Irritant
Formula:
C₁₄H₂₀O₂ |
MDL Number:
MFCD06246098 CAS Number: 883515-73-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...