5-(tert-Butyl)-2-ethoxybenzaldehyde

Supplier: Matrix Scientific

053615-500MG 053615-2.5G
101904-346EA 364.62 USD
101904-346 101904-348
5-(tert-Butyl)-2-ethoxybenzaldehyde
5-(tert-Butyl)-2-ethoxybenzaldehyde

Hazard: Irritant

Formula: C₁₃H₁₈O₂
MDL Number: MFCD02380389
CAS Number: 681443-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR