3-Chloro-6-(cyclopropylmethoxy)-2,4-dimethylbenzaldehyde
Supplier: Matrix Scientific
053612-500MG
053612-2.5G
101904-334EA
364.62
USD
101904-334
101904-336
3-Chloro-6-(cyclopropylmethoxy)-2,4-dimethylbenzaldehyde
3-Chloro-6-(cyclopropylmethoxy)-2,4-dimethylbenzaldehyde
Hazard: Irritant
Formula:
C₁₃H₁₅ClO₂ |
MDL Number:
MFCD06246138 CAS Number: 883519-38-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...