3-Chloro-2,4-dimethyl-6-propoxybenzaldehyde

Supplier: Matrix Scientific

053611-500MG 053611-2.5G
101904-330EA 364.62 USD
101904-330 101904-332
3-Chloro-2,4-dimethyl-6-propoxybenzaldehyde
3-Chloro-2,4-dimethyl-6-propoxybenzaldehyde

Hazard: Irritant

Formula: C₁₂H₁₅ClO₂
MDL Number: MFCD06246167
CAS Number: 883520-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR