3-Chloro-6-ethoxy-2,4-dimethylbenzaldehyde
Supplier: Matrix Scientific
053610-500MG
053610-2.5G
101904-326EA
364.62
USD
101904-326
101904-328
3-Chloro-6-ethoxy-2,4-dimethylbenzaldehyde
3-Chloro-6-ethoxy-2,4-dimethylbenzaldehyde
Hazard: Irritant
Formula:
C₁₁H₁₃ClO₂ |
MDL Number:
MFCD06246174 CAS Number: 883521-32-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...